Product Details

SNP ID
rs74837062
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36219508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGCATGTATGTGTGATATATA[C/T]ATAACATGTTTTGTCCATTTATTTC
Phenotype
MIM: 118960 MIM: 603824
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CLTA PubMed Links

Gene Details

Gene
CLTA
Gene Name
clathrin light chain A
There are no transcripts associated with this gene.

Gene
GNE
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128227.2 Intron NP_001121699.1
NM_001190383.1 Intron NP_001177312.1
NM_001190384.1 Intron NP_001177313.1
NM_001190388.1 Intron NP_001177317.1
NM_005476.5 Intron NP_005467.1
XM_005251334.4 Intron XP_005251391.1
XM_017014167.1 Intron XP_016869656.1
XM_017014168.1 Intron XP_016869657.1

View Full Product Details