Product Details
- SNP ID
-
rs74372765
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:89642053 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCATTATGCCTCGGGTGTTTGACC[C/T]GGCTGGATATGGGATGGAGAGCGTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UNQ6494
PubMed Links
Gene Details
- Gene
- UNQ6494
- Gene Name
- uncharacterized LOC100129066
There are no transcripts associated with this gene.
View Full Product Details