Product Details
- SNP ID
-
rs77739970
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:96464399 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAAAAAAAGAAATCTAAAGTAAG[A/G]TTCTAATAAAAGAAAAGTGAGAGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HABP4
PubMed Links
Gene Details
- Gene
- HABP4
- Gene Name
- hyaluronan binding protein 4
View Full Product Details