Product Details

SNP ID
rs80076221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104617530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCAGAGGAGTGAATCTTGAG[G/A]ATGCTGGAAATGATTAATGAGTAAG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
OR13C9 PubMed Links

Gene Details

Gene
OR13C9
Gene Name
olfactory receptor family 13 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001956.1 675 Silent Mutation ATC,ATT I225I NP_001001956.1

View Full Product Details