Product Details

SNP ID
rs76739796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128730621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTCCACAGCCTGGGACCTGGT[A/G]GGAGACTAGGAGGCAGCCTCCATAG
Phenotype
MIM: 610714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
PKN3
Gene Name
protein kinase N3
There are no transcripts associated with this gene.

Gene
ZDHHC12
Gene Name
zinc finger DHHC-type containing 12
There are no transcripts associated with this gene.

Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006336.3 3257 UTR 3 NP_006327.2
XM_005251645.2 3257 UTR 3 XP_005251702.1
XM_005251646.2 3257 UTR 3 XP_005251703.1
XM_011518122.2 3257 UTR 3 XP_011516424.1
XM_017014185.1 3257 UTR 3 XP_016869674.1
XM_017014186.1 3257 UTR 3 XP_016869675.1
XM_017014187.1 3257 UTR 3 XP_016869676.1
XM_017014188.1 3257 UTR 3 XP_016869677.1
XM_017014189.1 3257 UTR 3 XP_016869678.1
XM_017014190.1 3257 UTR 3 XP_016869679.1

View Full Product Details