Product Details

SNP ID
rs75629094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:34110816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACAGGGCCGCACGTCATCCTC[C/T]ACTTCAGGTGAATTTGTCACTGCAA
Phenotype
MIM: 606745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARD3 PubMed Links

Gene Details

Gene
PARD3
Gene Name
par-3 family cell polarity regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184785.1 4577 UTR 3 NP_001171714.1
NM_001184786.1 4577 UTR 3 NP_001171715.1
NM_001184787.1 4577 UTR 3 NP_001171716.1
NM_001184788.1 4577 UTR 3 NP_001171717.1
NM_001184789.1 4577 UTR 3 NP_001171718.1
NM_001184790.1 4577 UTR 3 NP_001171719.1
NM_001184791.1 4577 UTR 3 NP_001171720.1
NM_001184792.1 4577 Intron NP_001171721.1
NM_001184793.1 4577 Intron NP_001171722.1
NM_001184794.1 4577 Intron NP_001171723.1
NM_019619.3 4577 UTR 3 NP_062565.2
XM_005252528.4 4577 UTR 3 XP_005252585.1
XM_005252530.4 4577 UTR 3 XP_005252587.1
XM_005252531.4 4577 UTR 3 XP_005252588.1
XM_005252532.4 4577 UTR 3 XP_005252589.1
XM_005252534.4 4577 Intron XP_005252591.1
XM_005252535.4 4577 Intron XP_005252592.1
XM_005252536.4 4577 Intron XP_005252593.1
XM_011519569.2 4577 UTR 3 XP_011517871.1
XM_011519570.2 4577 UTR 3 XP_011517872.1
XM_011519571.2 4577 UTR 3 XP_011517873.1
XM_011519572.2 4577 UTR 3 XP_011517874.1
XM_011519573.2 4577 UTR 3 XP_011517875.1
XM_011519575.2 4577 UTR 3 XP_011517877.1
XM_011519576.2 4577 UTR 3 XP_011517878.1
XM_011519577.2 4577 UTR 3 XP_011517879.1
XM_011519578.2 4577 UTR 3 XP_011517880.1
XM_011519579.2 4577 UTR 3 XP_011517881.1
XM_011519581.2 4577 Intron XP_011517883.1
XM_011519583.2 4577 Intron XP_011517885.1
XM_011519584.2 4577 Intron XP_011517886.1
XM_017016430.1 4577 Intron XP_016871919.1

View Full Product Details