Product Details

SNP ID
rs76889310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14286866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTACCCGAACAATTAACGATAAG[A/G]AAGGAAGCCGAGTAAACACAGAAAG
Phenotype
MIM: 600098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRAS2 PubMed Links

Gene Details

Gene
RRAS2
Gene Name
related RAS viral (r-ras) oncogene homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102669.2 Intron NP_001096139.1
NM_001177314.1 Intron NP_001170785.1
NM_001177315.1 Intron NP_001170786.1
NM_012250.5 Intron NP_036382.2
XM_017017363.1 Intron XP_016872852.1

View Full Product Details