Product Details

SNP ID
rs76765539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2300742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGCCCGCCTCACCAGCTCCAG[A/G]GAGGTCAAGGTTGGAGAGAGACAAT
Phenotype
MIM: 611033 MIM: 603853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf21 PubMed Links

Gene Details

Gene
C11orf21
Gene Name
chromosome 11 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520034.1 185 Missense Mutation CCT,TCT P45S XP_011518336.1
Gene
TSPAN32
Gene Name
tetraspanin 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139022.2 185 Intron NP_620591.3
XM_005252719.4 185 Intron XP_005252776.1
XM_005252720.4 185 Intron XP_005252777.1
XM_011519815.2 185 Intron XP_011518117.2
XM_011519818.2 185 Intron XP_011518120.2
XM_011519819.1 185 Intron XP_011518121.1
XM_011519822.1 185 Intron XP_011518124.1
XM_011519828.2 185 Intron XP_011518130.1
XM_011519829.2 185 Intron XP_011518131.2
XM_017017064.1 185 Intron XP_016872553.1
XM_017017065.1 185 Intron XP_016872554.1
XM_017017066.1 185 Intron XP_016872555.1
XM_017017067.1 185 Intron XP_016872556.1
XM_017017068.1 185 Intron XP_016872557.1
XM_017017069.1 185 Intron XP_016872558.1
XM_017017070.1 185 Intron XP_016872559.1
XM_017017071.1 185 Intron XP_016872560.1

View Full Product Details