Product Details

SNP ID
rs75897392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33858763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAAACAGTGATTCATCAGCATT[A/G]CTTTAAATAATTGTTTGGTTAAAAG
Phenotype
MIM: 180385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMO2 PubMed Links

Gene Details

Gene
LMO2
Gene Name
LIM domain only 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142315.1 1861 UTR 3 NP_001135787.1
NM_001142316.1 1861 UTR 3 NP_001135788.1
NM_005574.3 1861 UTR 3 NP_005565.2
XM_005252921.2 1861 UTR 3 XP_005252978.1
XM_017017727.1 1861 UTR 3 XP_016873216.1
XM_017017728.1 1861 UTR 3 XP_016873217.1
XM_017017729.1 1861 UTR 3 XP_016873218.1
XM_017017730.1 1861 UTR 3 XP_016873219.1
XM_017017731.1 1861 UTR 3 XP_016873220.1
XM_017017732.1 1861 UTR 3 XP_016873221.1
XM_017017733.1 1861 UTR 3 XP_016873222.1

View Full Product Details