Product Details

SNP ID
rs77436432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46936699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGTTTGTGTGACACCTCCTCCA[A/G]CTCCCGTCCCCCTTCCGCCGCGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf49 PubMed Links

Gene Details

Gene
C11orf49
Gene Name
chromosome 11 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003676.2 2 UTR 5 NP_001003676.1
NM_001003677.2 2 UTR 5 NP_001003677.1
NM_001003678.2 2 UTR 5 NP_001003678.1
NM_001278222.1 2 UTR 5 NP_001265151.1
NM_024113.4 2 UTR 5 NP_077018.1
XM_006718315.1 2 UTR 5 XP_006718378.1
XM_011520364.2 2 UTR 5 XP_011518666.1
XM_011520365.2 2 UTR 5 XP_011518667.1
XM_017018276.1 2 UTR 5 XP_016873765.1
XM_017018277.1 2 Intron XP_016873766.1
XM_017018278.1 2 UTR 5 XP_016873767.1
XM_017018279.1 2 UTR 5 XP_016873768.1
XM_017018280.1 2 Intron XP_016873769.1
XM_017018281.1 2 UTR 5 XP_016873770.1
XM_017018282.1 2 UTR 5 XP_016873771.1

View Full Product Details