Product Details

SNP ID
rs77813967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8612081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTTTTATTTGTTCTTGAATGT[A/G]TAGGTTCTTTTAAAATCAGCTATAC
Phenotype
MIM: 612000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM66 PubMed Links

Gene Details

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 10563 UTR 3 NP_055633.1
XM_006718397.2 10563 UTR 3 XP_006718460.1
XM_006718398.3 10563 UTR 3 XP_006718461.1
XM_011520504.1 10563 UTR 3 XP_011518806.1
XM_011520507.2 10563 UTR 3 XP_011518809.1
XM_011520508.1 10563 UTR 3 XP_011518810.1
XM_011520509.1 10563 UTR 3 XP_011518811.1
XM_011520510.1 10563 UTR 3 XP_011518812.1
XM_011520511.1 10563 UTR 3 XP_011518813.1
XM_011520512.1 10563 UTR 3 XP_011518814.1
XM_011520513.1 10563 UTR 3 XP_011518815.1
XM_011520514.2 10563 UTR 3 XP_011518816.1
XM_011520515.1 10563 UTR 3 XP_011518817.1
XM_011520516.1 10563 UTR 3 XP_011518818.1
XM_011520517.2 10563 UTR 3 XP_011518819.1
XM_011520518.1 10563 UTR 3 XP_011518820.1
XM_011520519.1 10563 UTR 3 XP_011518821.1
XM_011520522.2 10563 UTR 3 XP_011518824.1
XM_011520523.2 10563 UTR 3 XP_011518825.1
XM_011520524.1 10563 Intron XP_011518826.1
XM_011520525.1 10563 Intron XP_011518827.1
XM_011520526.2 10563 UTR 3 XP_011518828.1
XM_011520527.1 10563 UTR 3 XP_011518829.1
XM_017018629.1 10563 UTR 3 XP_016874118.1
XM_017018630.1 10563 UTR 3 XP_016874119.1
XM_017018631.1 10563 UTR 3 XP_016874120.1

View Full Product Details