Product Details

SNP ID
rs79020237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:125197489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCATCAGCAAGCCATAGGCCA[C/T]GGACCAAATTTGGCCCTCTACTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM132B PubMed Links

Gene Details

Gene
TMEM132B
Gene Name
transmembrane protein 132B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286219.1 Intron NP_001273148.1
NM_052907.3 Intron NP_443139.2
XM_011537852.2 Intron XP_011536154.1
XM_011537854.2 Intron XP_011536156.1
XM_017018767.1 Intron XP_016874256.1

View Full Product Details