Product Details

SNP ID
rs74920894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130341993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACGTATATGGTGATGCCACTCA[A/C]CAAGAATGAAATTATGGGACATGAG
Phenotype
MIM: 605571
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PIWIL1 PubMed Links

Gene Details

Gene
PIWIL1
Gene Name
piwi like RNA-mediated gene silencing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190971.1 Intron NP_001177900.1
NM_004764.4 Intron NP_004755.2
XM_011539002.2 Intron XP_011537304.1
XM_011539003.2 Intron XP_011537305.1
XM_011539004.2 Intron XP_011537306.1
XM_011539005.1 Intron XP_011537307.1
XM_011539006.2 Intron XP_011537308.1
XM_017020229.1 Intron XP_016875718.1

View Full Product Details