Product Details

SNP ID
rs78013033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120095198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCAAACCATTGAAAAGTGGTGT[G/T]TGTTTGACAGGAATTTCACATCAGG
Phenotype
MIM: 617002 MIM: 604199
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BICDL1 PubMed Links

Gene Details

Gene
BICDL1
Gene Name
BICD family like cargo adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207311.2 2619 Intron NP_997194.2
XM_005253991.4 2619 Intron XP_005254048.1
XM_006719694.3 2619 Intron XP_006719757.1
XM_006719696.3 2619 Intron XP_006719759.1
XM_006719697.3 2619 Intron XP_006719760.1
XM_011538999.2 2619 Intron XP_011537301.1
XM_011539000.2 2619 Intron XP_011537302.1
XM_011539001.2 2619 Intron XP_011537303.1
Gene
RAB35
Gene Name
RAB35, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167606.1 2619 UTR 3 NP_001161078.1
NM_006861.6 2619 UTR 3 NP_006852.1
XM_011537779.1 2619 UTR 3 XP_011536081.1
XM_017018723.1 2619 UTR 3 XP_016874212.1

View Full Product Details