Product Details

SNP ID
rs75498472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:87675700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATCCCAATGAGTTTGTCAATTA[C/T]ACTGGGGCTTCAATTTTGCATCTAG
Phenotype
MIM: 609680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4500HG PubMed Links

Gene Details

Gene
MIR4500HG
Gene Name
MIR4500 host gene
There are no transcripts associated with this gene.

Gene
SLITRK5
Gene Name
SLIT and NTRK like family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015567.1 1202 Silent Mutation TAC,TAT Y104Y NP_056382.1
XM_005254038.4 1202 Silent Mutation TAC,TAT Y104Y XP_005254095.1
XM_005254039.2 1202 Silent Mutation TAC,TAT Y104Y XP_005254096.1

View Full Product Details