Product Details
- SNP ID
-
rs76921545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:106491379 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTGTTTTTCCCAGAAGTAGCTG[C/T]CCAATTTGTTTTTTTAAGCGCTGAG
- Phenotype
-
MIM: 600527
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EFNB2
PubMed Links
Gene Details
- Gene
- EFNB2
- Gene Name
- ephrin B2
View Full Product Details