Product Details

SNP ID
rs75385297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39603630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGGAGGGTCCCAGGGCATGCT[C/T]GCTCTAAGAGGAAGGTTGCCCAAAA
Phenotype
MIM: 615795 MIM: 188060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSIP1 PubMed Links

Gene Details

Gene
FSIP1
Gene Name
fibrous sheath interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324338.1 Intron NP_001311267.1
NM_152597.4 Intron NP_689810.3
XM_011521305.2 Intron XP_011519607.1
XM_011521306.1 Intron XP_011519608.1
XM_011521307.2 Intron XP_011519609.1
XM_011521309.2 Intron XP_011519611.1
XM_011521311.2 Intron XP_011519613.1
XM_017021972.1 Intron XP_016877461.1
Gene
THBS1
Gene Name
thrombospondin 1
There are no transcripts associated with this gene.

View Full Product Details