Product Details

SNP ID
rs77515052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44441053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTCATGAGTGGGACTCTATCA[A/G]TAATCTTGCTCCTCTAGGCCAGACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDSPL2 PubMed Links

Gene Details

Gene
CTDSPL2
Gene Name
CTD small phosphatase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016396.2 Intron NP_057480.2
XM_005254441.2 Intron XP_005254498.1
XM_006720565.2 Intron XP_006720628.1
XM_011521665.2 Intron XP_011519967.1
XM_017022306.1 Intron XP_016877795.1
XM_017022307.1 Intron XP_016877796.1

View Full Product Details