Product Details

SNP ID
rs79340306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55185705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCACTTTAAACCAATCCAATG[C/T]TTTTCAAATTTTACTATAAAGAGAA
Phenotype
MIM: 613262
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSL24D1 PubMed Links

Gene Details

Gene
RSL24D1
Gene Name
ribosomal L24 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016304.2 Intron NP_057388.1

View Full Product Details