Product Details

SNP ID
rs80028556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:57954276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTCATTTAATAATAATTGTTG[C/G]CCAATATTAGAAACTGAATGATGTC
Phenotype
MIM: 603687
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALDH1A2 PubMed Links

Gene Details

Gene
ALDH1A2
Gene Name
aldehyde dehydrogenase 1 family member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206897.1 2751 UTR 3 NP_001193826.1
NM_003888.3 2751 UTR 3 NP_003879.2
NM_170696.2 2751 UTR 3 NP_733797.1
NM_170697.2 2751 UTR 3 NP_733798.1

View Full Product Details