Product Details

SNP ID
rs75763893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63279621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAAAAGACAGAGTCTCTGGCTT[C/T]ATGGAGCTTATATTATAGTAGGAAG
Phenotype
MIM: 607630
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APH1B PubMed Links

Gene Details

Gene
APH1B
Gene Name
aph-1 homolog B, gamma-secretase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145646.1 Intron NP_001139118.1
NM_031301.3 Intron NP_112591.2
XM_011522105.2 Intron XP_011520407.1

View Full Product Details