Product Details

SNP ID
rs77625823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64170359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTTTCTGGAAGGAGTCCAATT[C/G]AGAAGATAATTTCAGACCTTTCATT
Phenotype
MIM: 606274 MIM: 123841
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
There are no transcripts associated with this gene.

View Full Product Details