Product Details

SNP ID
rs78112942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74449991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCGGGAAGAGGAGTCAGTCCC[A/G]GGCATTGGGGCACTGCCTGCTACGG
Phenotype
MIM: 609748
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBL7 PubMed Links

Gene Details

Gene
UBL7
Gene Name
ubiquitin like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286739.1 719 Silent Mutation CCC,CCT P203P NP_001273668.1
NM_001286740.1 719 Silent Mutation CCC,CCT P203P NP_001273669.1
NM_001286741.1 719 Silent Mutation CCC,CCT P243P NP_001273670.1
NM_001286742.1 719 Silent Mutation CCC,CCT P243P NP_001273671.1
NM_032907.4 719 Silent Mutation CCC,CCT P203P NP_116296.1
NM_201265.1 719 Silent Mutation CCC,CCT P203P NP_957717.1
XM_011522124.2 719 Silent Mutation CCC,CCT P243P XP_011520426.1

View Full Product Details