Product Details

SNP ID
rs79037950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78169633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATATAATGAGAGATATAATTTTT[A/G]TTAATAAGACAAAGGTAATATATTG
Phenotype
MIM: 614362 MIM: 601149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSBG1 PubMed Links

Gene Details

Gene
ACSBG1
Gene Name
acyl-CoA synthetase bubblegum family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199377.1 1771 Intron NP_001186306.1
NM_015162.4 1771 Intron NP_055977.3
XM_011521390.2 1771 Intron XP_011519692.1
XM_011521391.2 1771 Intron XP_011519693.2
XM_017022025.1 1771 Intron XP_016877514.1
XM_017022026.1 1771 Intron XP_016877515.1
Gene
IDH3A
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005530.2 1771 UTR 3 NP_005521.1
XM_005254336.2 1771 UTR 3 XP_005254393.1
XM_017022118.1 1771 UTR 3 XP_016877607.1
XM_017022119.1 1771 UTR 3 XP_016877608.1
XM_017022120.1 1771 UTR 3 XP_016877609.1

View Full Product Details