Product Details

SNP ID
rs79353570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34858842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAAGTCAACAAAGAAAAAAAT[C/T]GCCTATTAGGGAAGTGGTGCTCAAA
Phenotype
MIM: 610548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQR PubMed Links

Gene Details

Gene
AQR
Gene Name
aquarius intron-binding spliceosomal factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014691.2 Intron NP_055506.1

View Full Product Details