Product Details

SNP ID
rs79171269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84687594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAAACACTTAAACAAAAGCACT[C/T]AGAAACAAAGATGCTATACTTTGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC11A PubMed Links

Gene Details

Gene
SEC11A
Gene Name
SEC11 homolog A, signal peptidase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271918.1 Intron NP_001258847.1
NM_001271919.1 Intron NP_001258848.1
NM_001271920.1 Intron NP_001258849.1
NM_001271921.1 Intron NP_001258850.1
NM_001271922.1 Intron NP_001258851.1
NM_014300.3 Intron NP_055115.1

View Full Product Details