Product Details

SNP ID
rs75494113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31120846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTGGGAAAGACGAGAATGTTA[C/T]TCGGCAGTTGCTATAGCCAAGCACC
Phenotype
MIM: 614901 MIM: 609912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCKDK PubMed Links

Gene Details

Gene
BCKDK
Gene Name
branched chain ketoacid dehydrogenase kinase
There are no transcripts associated with this gene.

Gene
KAT8
Gene Name
lysine acetyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032188.2 Intron NP_115564.2
NM_182958.2 Intron NP_892003.2
XM_011545970.1 Intron XP_011544272.1
XM_011545971.1 Intron XP_011544273.1

View Full Product Details