Product Details

SNP ID
rs79473231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68854661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCATGTGGATATCCAGTTGTTCC[A/C]ACCTCATTTGGTAAAGGAATTTTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TANGO6 PubMed Links

Gene Details

Gene
TANGO6
Gene Name
transport and golgi organization 6 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024562.1 Intron NP_078838.1
XM_011523327.2 Intron XP_011521629.1

View Full Product Details