Product Details

SNP ID
rs79088174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89660463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGGTGGGGGAGGAAGGTGGAC[A/G]TGAAGGGAAAACAGCAGAGCAAGAG
Phenotype
MIM: 164010 MIM: 615409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHMP1A PubMed Links

Gene Details

Gene
CHMP1A
Gene Name
charged multivesicular body protein 1A
There are no transcripts associated with this gene.

Gene
SPATA33
Gene Name
spermatogenesis associated 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271907.1 Intron NP_001258836.1
NM_001271908.1 Intron NP_001258837.1
NM_001271909.1 Intron NP_001258838.1
NM_001271910.1 Intron NP_001258839.1
NM_153025.2 Intron NP_694570.1
XM_006721133.2 Intron XP_006721196.1

View Full Product Details