Product Details
- SNP ID
-
rs76688449
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:90029074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTTGTCCCAGCATGTGAGCTGCTG[A/C]CCGTCTCCCTGTCCGCTTCCCATTC
- Phenotype
-
MIM: 605178
MIM: 605179
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DBNDD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3785184] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DBNDD1
- Gene Name
- dysbindin domain containing 1
There are no transcripts associated with this gene.
- Gene
- GAS8
- Gene Name
- growth arrest specific 8
- Gene
- GAS8-AS1
- Gene Name
- GAS8 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details