Product Details

SNP ID
rs76948795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82944529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGCATGTTGGGTGTTGACTCC[A/G]AGAAGGTGTTCAGAAAACCTCTCTG
Phenotype
MIM: 615337 MIM: 604649
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GNTL1 PubMed Links

Gene Details

Gene
B3GNTL1
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009905.2 Intron NP_001009905.1
NM_001320742.1 Intron NP_001307671.1
NM_001320743.1 Intron NP_001307672.1
XM_006722272.3 Intron XP_006722335.1
XM_006722274.2 Intron XP_006722337.2
XM_011523532.2 Intron XP_011521834.1
XM_011523535.1 Intron XP_011521837.1
XM_017024205.1 Intron XP_016879694.1
XM_017024206.1 Intron XP_016879695.1
XM_017024207.1 Intron XP_016879696.1
Gene
LOC105371944
Gene Name
uncharacterized LOC105371944
There are no transcripts associated with this gene.

Gene
TBCD
Gene Name
tubulin folding cofactor D
There are no transcripts associated with this gene.

View Full Product Details