Product Details

SNP ID
rs77747256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:511934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATTCACAGGAGACATGCCCTAT[C/T]TTCCACAACAGGCAGCCTCGCCACC
Phenotype
MIM: 615850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPS53 PubMed Links

Gene Details

Gene
VPS53
Gene Name
VPS53, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128159.2 9886 UTR 3 NP_001121631.1
NM_018289.3 9886 Intron NP_060759.2
XM_011523953.2 9886 Intron XP_011522255.1
XM_017024817.1 9886 UTR 3 XP_016880306.1
XM_017024818.1 9886 Intron XP_016880307.1

View Full Product Details