Product Details

SNP ID
rs74775774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:20881587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATATTAAAATATAAAAAAGATT[G/T]TGAATTTGTTTTAAAAATTAAGTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC144NL PubMed Links

Gene Details

Gene
CCDC144NL
Gene Name
coiled-coil domain containing 144 family, N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004306.2 Intron NP_001004306.1
XM_011523818.2 Intron XP_011522120.1
XM_011523819.2 Intron XP_011522121.1
XM_011523821.1 Intron XP_011522123.1
Gene
CCDC144NL-AS1
Gene Name
CCDC144NL antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details