Product Details
- SNP ID
-
rs76178720
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:9253873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAGAAATCAAGAACAACAACTGA[C/T]TGGGAAGAAGAAAACACACATCTGG
- Phenotype
-
MIM: 601614
MIM: 604203
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTN1
PubMed Links
Gene Details
- Gene
- NTN1
- Gene Name
- netrin 1
There are no transcripts associated with this gene.
- Gene
- STX8
- Gene Name
- syntaxin 8
View Full Product Details