Product Details

SNP ID
rs74628253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40126019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTATACCTTCTCTTCTCTATTTT[G/T]TTTAAGGAAAACTGTTCAGATTAAT
Phenotype
MIM: 614801
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MSL1 PubMed Links

Gene Details

Gene
MSL1
Gene Name
male specific lethal 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012241.1 Intron NP_001012241.1
XM_005257298.4 Intron XP_005257355.1
XM_005257299.4 Intron XP_005257356.3

View Full Product Details