Product Details

SNP ID
rs76774458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40129482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCAAAAGGGACCCAGCACCCA[C/T]CCCAAGGAGAAAGCCTTCTCAAGTG
Phenotype
MIM: 614801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSL1 PubMed Links

Gene Details

Gene
MSL1
Gene Name
male specific lethal 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012241.1 782 Silent Mutation CAC,CAT H147H NP_001012241.1
XM_005257298.4 782 Silent Mutation CAC,CAT H410H XP_005257355.1
XM_005257299.4 782 Silent Mutation CAC,CAT H410H XP_005257356.3

View Full Product Details