Product Details

SNP ID
rs74878401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5142717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATTAAAGATCCCTCTCTTTTG[C/T]ATTGTTTTATCATAGAAACCTTAAA
Phenotype
MIM: 616427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAIN1 PubMed Links

Gene Details

Gene
AKAIN1
Gene Name
A kinase (PRKA) anchor inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145194.1 Intron NP_001138666.1
XM_011525728.2 Intron XP_011524030.1

View Full Product Details