Product Details

SNP ID
rs76649895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9915909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTACGTAGTTGTAGCTGGGGGA[A/G]AACCTTTGAGGAATTATGAAGAGTC
Phenotype
MIM: 605703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAPA PubMed Links

Gene Details

Gene
VAPA
Gene Name
VAMP associated protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003574.5 Intron NP_003565.4
NM_194434.2 Intron NP_919415.2
XM_011525769.2 Intron XP_011524071.1
XM_017026079.1 Intron XP_016881568.1

View Full Product Details