Product Details

SNP ID
rs77505124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35253233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGTTAGGCATTTCAAGGAAATA[C/T]CTACCATTCTTTTTGGAGAAGACTT
Phenotype
MIM: 609601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF397 PubMed Links

Gene Details

Gene
ZNF397
Gene Name
zinc finger protein 397
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135178.2 2119 Intron NP_001128650.1
NM_032347.2 2119 Intron NP_115723.1
XM_006722558.2 2119 UTR 3 XP_006722621.1
XM_011526229.2 2119 UTR 3 XP_011524531.1
XM_011526230.2 2119 UTR 3 XP_011524532.1
XM_011526231.2 2119 UTR 3 XP_011524533.1
XM_017026042.1 2119 UTR 3 XP_016881531.1
Gene
ZSCAN30
Gene Name
zinc finger and SCAN domain containing 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112734.3 2119 UTR 3 NP_001106205.1
NM_001166012.2 2119 UTR 3 NP_001159484.1
NM_001288711.1 2119 UTR 3 NP_001275640.1
XM_005258183.4 2119 UTR 3 XP_005258240.1
XM_006722371.3 2119 UTR 3 XP_006722434.1
XM_011525789.2 2119 UTR 3 XP_011524091.1
XM_017025515.1 2119 UTR 3 XP_016881004.1
XM_017025516.1 2119 UTR 3 XP_016881005.1
XM_017025517.1 2119 UTR 3 XP_016881006.1
XM_017025518.1 2119 UTR 3 XP_016881007.1
XM_017025519.1 2119 UTR 3 XP_016881008.1
XM_017025520.1 2119 UTR 3 XP_016881009.1
XM_017025521.1 2119 UTR 3 XP_016881010.1
XM_017025522.1 2119 UTR 3 XP_016881011.1

View Full Product Details