Product Details

SNP ID
rs76140888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39515263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTCCTCGGCGCGGACTTCAAC[C/T]TCAGTCCGGGCTTCTACCCCGACCC
Phenotype
MIM: 602768 MIM: 607919
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
There are no transcripts associated with this gene.

Gene
SELV
Gene Name
selenoprotein V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182704.1 151 Silent Mutation ACC,ACT T17T NP_874363.1

View Full Product Details