Product Details
- SNP ID
-
rs79934495
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40075880 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACATTCAAAGGGTTTCTCTCCAG[C/T]ATGAATACTCTGATGCTGAATAAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF780A
PubMed Links
Gene Details
- Gene
- ZNF780A
- Gene Name
- zinc finger protein 780A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010880.2 |
713 |
Missense Mutation |
ACT,GCT |
T188A |
NP_001010880.2 |
NM_001142577.1 |
713 |
Missense Mutation |
ACT,GCT |
T189A |
NP_001136049.1 |
NM_001142578.1 |
713 |
Missense Mutation |
ACT,GCT |
T188A |
NP_001136050.1 |
NM_001142579.1 |
713 |
Intron |
|
|
NP_001136051.1 |
XM_005258773.2 |
713 |
Missense Mutation |
ACT,GCT |
T189A |
XP_005258830.1 |
XM_006723150.3 |
713 |
Missense Mutation |
ACT,GCT |
T189A |
XP_006723213.1 |
XM_011526771.2 |
713 |
Missense Mutation |
ACT,GCT |
T189A |
XP_011525073.1 |
XM_011526772.2 |
713 |
Missense Mutation |
ACT,GCT |
T189A |
XP_011525074.1 |
XM_017026611.1 |
713 |
Missense Mutation |
ACT,GCT |
T205A |
XP_016882100.1 |
XM_017026612.1 |
713 |
Missense Mutation |
ACT,GCT |
T205A |
XP_016882101.1 |
XM_017026613.1 |
713 |
Missense Mutation |
ACT,GCT |
T205A |
XP_016882102.1 |
XM_017026614.1 |
713 |
Missense Mutation |
ACT,GCT |
T205A |
XP_016882103.1 |
XM_017026615.1 |
713 |
Missense Mutation |
ACT,GCT |
T205A |
XP_016882104.1 |
XM_017026616.1 |
713 |
Missense Mutation |
ACT,GCT |
T204A |
XP_016882105.1 |
XM_017026617.1 |
713 |
Missense Mutation |
ACT,GCT |
T188A |
XP_016882106.1 |
XM_017026618.1 |
713 |
Missense Mutation |
ACT,GCT |
T188A |
XP_016882107.1 |
XM_017026619.1 |
713 |
Missense Mutation |
ACT,GCT |
T188A |
XP_016882108.1 |
View Full Product Details