Product Details
- SNP ID
-
rs74461539
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43872526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCATAATAGTGACAACAGTAAAAA[C/T]GTGCCATGGCTAAAGGCTTTCCCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF404
PubMed Links
Gene Details
- Gene
- ZNF404
- Gene Name
- zinc finger protein 404
View Full Product Details