Product Details
- SNP ID
-
rs78841585
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43873839 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACACTCATATGATTTCTCTCTTGT[A/G]TTATTTCTCTTATGTAGAGGAAGGG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF404
PubMed Links
Gene Details
- Gene
- ZNF404
- Gene Name
- zinc finger protein 404
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033719.2 |
375 |
Silent Mutation |
AAC,AAT |
N125N |
NP_001028891.2 |
View Full Product Details