Product Details

SNP ID
rs79968982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45181191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTGTCGTGGGATAGGTTTTGC[C/T]TTCTTTTACTGTCTCCATCCTACTT
Phenotype
MIM: 609762 MIM: 610396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S3 PubMed Links

Gene Details

Gene
BLOC1S3
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_212550.4 1991 UTR 3 NP_997715.1
Gene
TRAPPC6A
Gene Name
trafficking protein particle complex 6A
There are no transcripts associated with this gene.

View Full Product Details