Product Details

SNP ID
rs78449793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52308261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACAAGCACCTAGTAAATTCTAG[A/G]AAAGGAGTGAAGGAAGGAAAACGTT
Phenotype
MIM: 613910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF480 PubMed Links

Gene Details

Gene
ZNF480
Gene Name
zinc finger protein 480
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297624.1 Intron NP_001284553.1
NM_001297625.1 Intron NP_001284554.1
NM_144684.3 Intron NP_653285.2
XM_011526465.2 Intron XP_011524767.1

View Full Product Details