Product Details

SNP ID
rs74621876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53015658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATCTCCCTTGGATGCCAGTGGG[C/G]AAAGATTCTGCAACTACGGAGGGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERVV-1 PubMed Links

Gene Details

Gene
ERVV-1
Gene Name
endogenous retrovirus group V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152473.2 Intron NP_689686.2
Gene
LOC107985332
Gene Name
uncharacterized LOC107985332
There are no transcripts associated with this gene.

View Full Product Details