Product Details

SNP ID
rs75870827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55529807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGCTGCCTCCAGGGGCGCCCCA[C/G]GTGCTCCCTGATGGCGATCACAGCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101401.2 988 Silent Mutation CTG,GTG L325V NP_001094871.2
XM_006723327.3 988 Silent Mutation CTG,GTG L325V XP_006723390.1
XM_011527227.2 988 Intron XP_011525529.1

View Full Product Details