Product Details

SNP ID
rs78345835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34592984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAAAAAGGACAGCTTGGGTAG[A/G]TAGGGATGGAGAAGTGTGTTAAGGT
Phenotype
MIM: 615063
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCGB1B2P PubMed Links

Gene Details

Gene
SCGB1B2P
Gene Name
secretoglobin family 1B member 2, pseudogene
There are no transcripts associated with this gene.

Gene
SCGB2B2
Gene Name
secretoglobin family 2B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025591.3 884 UTR 3 NP_001020762.1

View Full Product Details