Product Details

SNP ID
rs79272253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35907756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGGGCCTGGGCCTGGACAGGA[C/T]GGAGACCTGAGGAGGAAAAAGAAGG
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
There are no transcripts associated with this gene.

Gene
NFKBID
Gene Name
NFKB inhibitor delta
There are no transcripts associated with this gene.

Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173514.1 149 Intron NP_001166985.1
NM_001173515.1 149 Intron NP_001166986.1
NM_003332.3 149 Missense Mutation CAT,CGT H23R NP_003323.1
NM_198125.2 149 Missense Mutation CAT,CGT H23R NP_937758.1

View Full Product Details