Product Details

SNP ID
rs79651410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36225359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTCCTATTCTTCTGTTTTCTGT[A/C]GATTCCCTTCTTTTGGGTGACTGTT
Phenotype
MIM: 601505
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF146 PubMed Links

Gene Details

Gene
ZNF146
Gene Name
zinc finger protein 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099638.1 Intron NP_001093108.1
NM_001099639.1 Intron NP_001093109.1
NM_007145.2 Intron NP_009076.2
XM_005259214.2 Intron XP_005259271.1
XM_017027244.1 Intron XP_016882733.1
XM_017027245.1 Intron XP_016882734.1
XM_017027246.1 Intron XP_016882735.1
XM_017027247.1 Intron XP_016882736.1

View Full Product Details